Squared bossheads
Leverancier: BOCHEM
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
B08600121 B08599125
241-3172EA 25.4 EUR
241-3172 BOCHB08599125
Squared bossheads
Klemmen Statiefkoppelingen
Squared bossheads with hexagon socket screws.
  • Made from 18/10 stainless steel or aluminium (laboral)
  • Angle: 90°
Order Now
Specificaties
Environmentally Preferable