Micro pipettes, disposable, Wiretrol® II
Leverancier: DRUMMOND
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
5-000-2005 5-000-2010 5-000-2020 5-000-2050 5-000-2100 5-000-2200
DRUM5-000-2005EA 135 EUR
DRUM5-000-2005 DRUM5-000-2010 DRUM5-000-2020 DRUM5-000-2050 DRUM5-000-2100 DRUM5-000-2200
Micro pipettes, disposable, Wiretrol® II
Pipetten Micro-capillairen
Wiretrol® II micro pipettes provide the accuracy and convenience of the Wiretrol I but with dual graduations and a shorter plunger for non contact delivery.
  • Economical positive displacement micro pipette with fingertip control
  • Dual graduations for non contact delivery
  • Ideal for a wide variety of applications, including both high and low viscosity liquids, and corrosives