Cryoboxes
Leverancier: HEATHROW
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
HS2860T HS2860M HS2860P
HEATHS2860TEA 71.9 EUR
HEATHS2860T HEATHS2860M HEATHS2860P
Cryoboxes
Dozen Dozen voor cryobuisjes
Made of cardboard, white.
  • Write-on surface for descriptions or self numbering systems
  • Dividers are flat packed and comes in two different sizes (to be ordered separately)
For storage of large samples and 15 or 50 ml tubes.