Labelling tapes, Write-On™, SP Bel-Art
Leverancier: Bel-Art Products, a Part of SP
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
F134620300 F134820075 F134840050 F134800050 F134830100 F134830075 F134840100 F134820100 F134850050 F134820050 F134810100 F134810075 F134810050 F134800100 F134800075 F134830050 F134870075 F134900100 F134900075 F134900050 F134880100 F134880075 F134840075 F134870100 F134870050 F134860100 F134860075 F134860050 F134850100 F134850075 F134880050
BELAF134620300EA 58.7 EUR
BELAF134620300 817-0195 817-0200 817-0188 817-0199 817-0198 817-0202 817-0196 817-0203 817-0194 817-0193 817-0192 817-0191 817-0190 817-0189 817-0197 817-0210 817-0217 817-0216 817-0215 817-0214 817-0213 817-0201 817-0211 817-0209 817-0208 817-0207 817-0206 817-0205 817-0204 817-0212
Labelling tapes, Write-On™, SP Bel-Art
Tape
These paper labelling tapes adhere to any clean surface, including PTFE coated materials, and peel off without leaving a sticky residue. Pencil, ball-point pen or solvent ink marker remain legible under the most demanding laboratory conditions.
  • Coloured paper with rubber resin adhesive resists moisture and most solvents
  • Withstands a maximum temperature of +157 °C for 30 minutes or +80 °C for 24 hours; can be used down to −80 °C
  • Withstands steam autoclaving cycle at +121 °C and gamma irradiation between 13 and 45 kGy
  • Excellent for use with Write-On™ multi-roll label tape dispensers
Available in three lengths and in different colours.