Schepjes, met gebogen handvat, Sterileware®, SP Bel-Art
Leverancier: Bel-Art Products, a Part of SP
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
H369042006 H369022006
212-0712EA 108 EUR
212-0712 212-0711
Schepjes, met gebogen handvat, Sterileware®, SP Bel-Art
Scheppen
PS, white, sterile. These sampling scoops are designed for retrieving samples from a larger source.
  • Flat bottom keeps scoops upright when placed on a flat surface making them excellent for use as weigh boats
  • Made from durable, FDA grade PS
  • Safe for food, drug and cosmetic contact
  • Individually packaged, sterilised by gamma irradiation
  • Lot stamped for accurate lot tracing; shipped with a Certificate of Processing for Sterilisation
The curved handle design positions the hand further from the sample when compared to straight handle scoops. This reducesMore Product Information