Bottles with pump vaporiser
Leverancier: KOEHLER TECHNISCHE PRODUKTEN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
K-2314 K-2313 K-2312 K-2310 K-2311
215-6274EA 6.3 EUR
215-6274 215-6273 215-6272 215-6270 215-6271
Bottles with pump vaporiser
Flessen Flessen met verstuiver
Amber soda glass bottles with pump vaporiser and suitable for chromatography.
  • Pump volume 0,12 ml per stroke
  • Good chemical resistance
  • Amber coloured for light sensitive substances