Racks for test tubes
Leverancier: KOEHLER TECHNISCHE PRODUKTEN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
A-1022 A-1021 A-1020
212-8382EA 136 EUR
212-8382 212-8381 212-8380
Racks for test tubes
Rekken Tube Racks
Made of acrylic, clear.
These acrylic test tube racks, two tier is a stable, solid, hygienic, easily cleaned, physiologically inert. Recesses in bases.