Pipette Boxes, Square
Leverancier: Köhler Technische Produkte
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
612-2044EA 102 EUR
612-2044 612-2045 KOEHW-1150 612-2041 612-2042 612-2043
Pipette Boxes, Square
Dozen Pipetsterilisatiecontainers
Anodized aluminium boxes with silicone insert for the protective storage of pipettes.
  • With slip-on cover
  • Autoclavable up to 180 °C