Cleanroom tables, perf top
Leverancier: Metro International
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
CTP3036S CTP3048S CTP3060S CTP3072S
METOCTP3036SEA 3725 EUR
METOCTP3036S METOCTP3048S METOCTP3060S METOCTP3072S
Cleanroom tables, perf top
Meubilair Tafels
Cleanroom table in stainless steel 304 with electropolished finish and anodised satin connectors.
  • Height adjustable legs
  • Tables with overheads have 1880 mm high rear posts
  • Non-conductive
Bestelinformatie: Cantilever shelves are sold separately.