Cleaning system for controlled environment
Leverancier: FHP VILEDA SCS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
141928 141926 143774 143776
VILE141928EA 214 EUR
VILE141928 141-0937 VILE143774 VILE143776
Cleaning system for controlled environment
Moppen
These cleanroom cleaning trolleys are specially designed for use in sensitive areas and controlled environments. The cleaning trolleys are extremely functional, versatile and are characterised by superior ergonomic properties.
  • Small and compact
  • Heat and chemical resistant
  • With electrically conductive castors, the tread of the wheels is made from an hydrolysis resistant material
  • Fully autoclavable and suitable for gamma sterilisation
ISO 5 / GMP A + B / D FED209 class 100
Certificaten: ISO 5 / GMP A + B / D FED209 class 100