Reusable half masks, RD40, N7700 series
Leverancier: Honeywell Safety
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
N65770016 N65770015
NORHN65770016EA 67.5 EUR
NORHN65770016 NORHN65770015
Reusable half masks, RD40, N7700 series
Ademhalingsmaskers Herbruikbare maskers Halfgelaatsmaskers
Ergonomically designed reusable half mask made from a silicone body ensuring extra comfort.
  • Twin exhalation valves for extra breathing comfort
  • Four-point head harness
  • Filter Cartridge Connection Type RD40
Colour: Black; Weight: 151 g
Certificaten: EN 140