Staining plates
Leverancier: GLASWARENFABRIK KARL HECHT
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
40623010 40624010
HECH40623010EA 33.1 EUR
HECH40623010 HECH40624010
Staining plates
Druppelplaten
Glass spot plates with 10 or 12 numbered cavities. For flocculation tests.