Flow cups with fixed nozzle 'Ford'
Leverancier: Industrial Physics
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
VF2032 VF2030 VF2031 VF2033
SIMEVF2032EA 210 EUR
SIMEVF2032 SIMEVF2030 SIMEVF2031 SIMEVF2033
Flow cups with fixed nozzle 'Ford'
Viscosimeters Flow-cup viscometers
Titanium anodised aluminium flow cups for measurement of relative viscosity.
  • Each cup has an engraved unique serial number
  • Design of cup and orifice facilitate cleaning process
  • Deep well surrounding the top of the cup to catch any overflow
Certificaten: ASTM D1200
Leveringsinformatie: Supplied with plastic case and internal soft protective material.