Specific gravity cups, calibrated
Leverancier: Industrial Physics
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
VF2099 VF2100 VF2097 VF2098
614-0715EA 377 EUR
614-0715 SIMEVF2100 SIMEVF2097 SIMEVF2098
Specific gravity cups, calibrated
Pyknometers
Specific gravity cups for determining the specific gravity or density (or weight per gallon wpg) of coatings, pastes or similar liquids.
  • Density is defined as weight per unit volume at a specified temperature
  • Available in anodised aluminium or stainless steel
Certificaten: According to ISO 2811, DIN 53 217 and ASTM D 1475.
Leveringsinformatie: Supplied with a calibration certificate.