Bottle-top dispensers, genius²
Leverancier: VITLAB
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
1625507 1625503 1625504 1625505 1625508 1625506
VITL1625507EA 562 EUR
VITL1625507 VITL1625503 VITL1625504 VITL1625505 VITL1625508 VITL1625506
Bottle-top dispensers, genius²
Flesdispensers
Autoclavable at 121 °C with adjustable volume.
  • Direct displacement plungers with a fluoroplastic (PFA) seal that works like a windscreen wiper to prevent build-up of easily crystallizing media on the cylinder wall
  • Glass cylinder coated with a synthetic material that reduces risk of splashes if breakage should occur
  • Telescopic intake tube can easily be smoothly adjusted to the height of the bottle
The genius² bottle top dispensers are precision devices that offer many advantages. Since the materials in contact with the media (PTFE, PFA, FEP, borosilicate glass and platinum-iridium) are resistant to acids, solvents and bases, they can be applied almost universally. They are equipped with a patented recirculation valve that reduces reagent loss when ventilated. The simple-to-use calibration function helps to meet all of the requirements of testing-apparatus monitoring, with the minimum of downtime.
Certificaten: According to DIN EN 285 and labelled DE-M.
Leveringsinformatie: Supplied with three threaded adapters made from PP. Nominal volumes of 2,5 to 10 ml (screw coupling GL 32) with adapters GL 28, S 40 and GL 45. Nominal volumes of 25 to 100 ml (screw coupling GL 45) with adapters GL 32, GL 38 and S 40. Telescopic filling tube (200 to 350 mm), mounting tool, instruction manual, and quality certificate stating all test values.