Chromatography paper
Leverancier: VARIAN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
SGI0001 A120B12
VARISGI0001EA 786 EUR
VARISGI0001 VARIA120B12
Chromatography paper
Filters Filterpapier Chromatografiepapier
Chromatography paper is used in thin layer chromatography applications such as evaluating radioisotope purity, QC testing.
  • The porous paper is made of glass microfibers impregnated with silica gel
  • No interference from organic binders
  • Separates lipids and other non-polar compounds
  • Can easily be cut to convenient testing sizes, and can be imprinted
Bestelinformatie: Offers two kinds of paper such as SA (contains sodium salt) and SG (contains potassium salt).