Single channel pipettes, fixed volume, Ovation® F1 / F2
Leverancier: VISTALAB TECHNOLOGIES
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
1157-0005 1157-0010 1157-0020 1157-0025 1157-0050 1157-0100 1157-0200 1157-0250 1157-0300 1157-0500 1157-1000 1257-0001 1257-0002 1257-0005 1257-0010 1257-0020 1257-0025 1257-0050 1257-0100 1257-0200 1257-0250 1257-0300 1257-0500 1257-1000
VIST1157-0005EA 527 EUR
VIST1157-0005 VIST1157-0010 VIST1157-0020 VIST1157-0025 VIST1157-0050 VIST1157-0100 VIST1157-0200 VIST1157-0250 VIST1157-0300 VIST1157-0500 VIST1157-1000 VIST1257-0001 VIST1257-0002 VIST1257-0005 VIST1257-0010 VIST1257-0020 VIST1257-0025 VIST1257-0050 VIST1257-0100 VIST1257-0200 VIST1257-0250 VIST1257-0300 VIST1257-0500 VIST1257-1000
Single channel pipettes, fixed volume, Ovation® F1 / F2
Pipetten Micropipetten
These fixed volume pipettes are the ideal ergonomic choice where pipetting volumes never deviate from an exact protocol. Both single stroke and two stroke (with overblow) models are available in volume increments from 1 to 1000 µl.
  • Comfortable, ergonomic design
  • Low-force, form-fitting plunger allows the user to manually control the speed of aspirating and dispensing with minimal contact or thumb effort
  • No excessive force or jamming is required to ensure a good tip seal
  • Filtered nozzle won't let liquid into the body of the device
  • Low force tip ejection
  • Built-in microtube cap opener
Leveringsinformatie: Starter pack includes one 10 µl, one 100 µl, one 200 µl, one 1000 µl pipette, an Ovation pipette stand, and three racks of tips.