Multi-channel pipettes, electronic, variable volume, Ovation BioNatural™
Leverancier: VISTALAB TECHNOLOGIES
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
1060-0020 1060-0125 1060-0250 1060-0850 1160-1250 1160-0020 1160-0125 1160-0250 1060-0020L 1060-0125L 1060-0250L 1060-0850L 1160-0020L 1160-0125L 1160-0250L 1160-1250L
VIST1060-0020EA 3000 EUR
VIST1060-0020 VIST1060-0125 VIST1060-0250 VIST1060-0850 VIST1160-1250 VIST1160-0020 VIST1160-0125 VIST1160-0250 VIST1060-0020L VIST1060-0125L VIST1060-0250L VIST1060-0850L VIST1160-0020L VIST1160-0125L VIST1160-0250L VIST1160-1250L
Multi-channel pipettes, electronic, variable volume, Ovation BioNatural™
Pipetten Micropipetten
This pipette is an adjustable volume, air displacement, fully electronic motor-driven pipette intended to aspirate and dispense precise fluid volumes. 8 and 12-channel models, in both left and right-handed configurations, are available to cover liquid dispensing needs between 0,5 and 1250 µl. Ideal for handling repetitive and complex liquid handling tasks.
  • Ergonomic design reduces fatigue and repetitive stress injuries
  • Larger grip surface for more comfort during operation
  • Easy tip ejection with minimal thumb force
  • LCD indicates liquid handling functions,