Cleanroom doekjes, MiraWIPE®
Leverancier: FOAMTEC
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
HT5790S HT5790B HT5790 HT5790S-10ST HT5790S-25 HT5790SM
FOAMHT5790SEA 0 EUR
FOAMHT5790S FOAMHT5790B FOAMHT5790 FOAMHT5790S-10ST FOAMHT5790S-25 FOAMHT5790SM
Cleanroom doekjes, MiraWIPE®
Schoonmaakdoekjes Cleanroom doekjes Droge cleanroom doekjes
These microfibre cleaning cloth wipes are constructed from a continuous filament micro denier (super soft polyester/nylon textile) resulting in improved absorbency and contamination pick up compared to traditional polyester wipes.
  • Laser cut wide-sealed edge construction
  • Woven from an abrasion resistant fabric to minimise In-Use particles from the face of the wipe
  • Non symmetrical microfibre structure allows it to scrub microbial bio-films and sporicidal residues making it ideal for disinfection procedures
  • Available sterile and non sterile
Ideal for removing stubbornly adhered particles and stains in pharmaceutical and micro electronics cleaning applications.