Plugs, PTFE, BOLA
Leverancier: Bohlender
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
F705-04 F705-02
BOHLF705-04EA 74.4 EUR
BOHLF705-04 BOHLF705-02
Plugs, PTFE, BOLA
Fittingen en connectoren
Made of PTFE, white.
  • Universal chemical resistance
  • Temperature resistance: from –200 to +260 °C
For closing unused ports of miniature fittings.