Thermostatic mixing valve for eye showers
Leverancier: BROEN-LAB
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
17.140.009 17140039
118-0137EA 871 EUR
118-0137 BROE17140039
Thermostatic mixing valve for eye showers
Ventielen Terugslagventielen
Thermostatic mixing valve for eye showers designed for delivering tepid water (20 to 25 °C).
  • Fail-safe on hot side: if the cold water is interrupted the built-in anti-scalding device will automatically cease the flow of hot water
  • Fail-safe on cold side: in the hot water is interrupted the built-in intentional leak will allow cold water to flow at a sufficient flowrate to ensure operation
  • Incl. check valves on both hot and cold supply
Weight: Approx. 1,5 kg
Min. working pressure: 1 bar