Lab Markers, several colours
Leverancier: PRECISION DYNAMICS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
1400-13-PDC 1400-15-PDC 1400-16-PDC 1400-20-PDC
PRCI1400-13-PDCEA 0 EUR
PRCI1400-13-PDC PRCI1400-15-PDC PRCI1400-16-PDC PRCI1400-20-PDC
Lab Markers, several colours
Pennen Stiften voor laboratoria
Versatile, permanent markings write on plastic, metal, and porcelain labware.
  • Several colours
  • Unique ink has the ability to write on labware even when the surface is damp