Cytology brushes
Leverancier: DELTALAB
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
440151 440150
DELT440151EA 437 EUR
DELT440151 DELT440150
Cytology brushes
Borstels
These cytology brushes are ideal for cervical specimen collection, designed for taking cell samples without damaging them.
  • Blue ABS shaft
  • Includes a soft tip to provide more comfort for patients
  • Sterile model is sterilised by ethylene oxide
Opgelet: *De product(en) op deze pagina vallen buiten het toepassingsgebied van de EU-richtlijn 2019/904