Tongue depressor
Leverancier: DELTALAB
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
441142.1 441142.2
231-2645EA 58.5 EUR
231-2645 231-2646
Tongue depressor
Tongspatel
PS, cream. Tongue depressor with smooth rounded edges.
  • Odour- and taste-free
  • Maintains a high tensile strength, more flexible than wood depressors
  • Features a ribbed finger rest for easy handling
Can also be used for spreading balms and ointments, for stirring liquids or mixing medicine.
Opgelet: *De product(en) op deze pagina vallen buiten het toepassingsgebied van de EU-richtlijn 2019/904