Assemblies, NanoPort™, Upchurch Scientific®
Leverancier: UPCHURCH SCIENTIFIC
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
N-123-02 N-123-03 N-123-03X N-333 N-333-01
UPCHN-123-02EA 62.1 EUR
UPCHN-123-02 UPCHN-123-03 UPCHN-123-03X UPCHN-333 UPCHN-333-01
Assemblies, NanoPort™, Upchurch Scientific®
Fittingen en connectoren
NanoPort™ assemblies provide consistent fluid connections for chip-based analyses, connections will bond to a variety of substrate materials with the use of Loctite™.
  • All components are made of inert, biocompatible material
  • Unique design prevents adhesive contamination of the fluid path
These connections add no additional volume to the fluid path, virtually eliminating dead volume traditionally associated with chip-based fluid connections.
Bestelinformatie: Adhesive is not included, need to be ordered separately.