Cryoboxes with Lid
Leverancier: DELTALAB
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
M-600 M-604 M-606 M-607 M-610
DELTM-600EA 227 EUR
DELTM-600 DELTM-604 DELTM-606 DELTM-607 DELTM-610
Cryoboxes with Lid
Dozen Dozen voor cryobuisjes
Cryoboxes, cardboard, white, with pre-assembled compartments are designed for storage and preservation of frozen samples.
  • Covered by special white paper hand-stuck and hand-assembled for a better quality
  • Resist temperatures up to –100 °C
  • Alphanumeric identifications printed on each box
Suitable for the racks of freezer chests. Ideal for microtubes and cryovials.