Measuring cylinders, tall form, PTFE
Leverancier: Bohlender
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
BOLA
BOHLA164-20EA 757 EUR
BOHLA164-20 BOHLA164-12 BOHLA164-16
Measuring cylinders, tall form, PTFE
Cilinders Maatcilinders
PTFE, tall form.
  • Ideal for volume measurement of aggressive liquids
  • Translucent and non porous with spout and reinforcing ring
  • Hexagonal base prevents cylinder from rolling
  • Permanent recessed graduation
Ø = Internal diameter
Ø = Internal diameter