Cryo-Clear™ laser cryo labels
Leverancier: DIVERSIFIED BIOTECH
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
CLEAR-170 CLEAR-238
817-0078EA 204 EUR
817-0078 817-0079
Cryo-Clear™ laser cryo labels
Etiketten
These Cryo-Clear™ vinyl cryo labels are specially top-coated to accept laser printing and other markings.
  • Temperature resistant from −196 to 70 °C
  • Chemically inert, resist most organic solvents and caustic agents
  • Adhere to plastics, glass and metals without cracking, peeling or degrading
Cryo-Clear™ labels are made with a flexible, vinyl label material and permanent acrylic adhesive. This unique combination expands and contracts together as temperatures fluctuate resulting in a strong and sustained adhesive bond. They remain fully adhered down to −196°C (liquid and vapor phase nitrogen).