BioHPLC columns, DNAPac™ PA100
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
088759 088761 043010 043011 043018 088765
555-7434EA 8105.72 EUR
555-7434 555-7436 555-7271 555-7272 555-7273 555-7440
BioHPLC columns, DNAPac™ PA100
Kolommen voor chromatografie
These columns feature a strong anion exchange resin for high resolution analysis and purification of synthetic and modified oligonucleotides.