pH electrodes with single rod measuring chain, ProcessLine
Leverancier: WTW ONLINE
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
109233 109239
WTWO109233EA 317 EUR
WTWO109233 WTWO109239
pH electrodes with single rod measuring chain, ProcessLine
Elektroden pH-elektroden
The special design of the process line electrodes makes them close to optimal in their accuracy, stability, speed and longevity to liquid electrolyte electrodes. For the use in pipelines, they can be installed into the replacement fitting CHEMtrac 830 M.
  • Easy to maintain by easy measuring chain replacement