Capillary columns for dioxin and PCB analysis, TRACE™
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
26AF047P 26AF059P 26AF154P 26AJ148P
555-9075EA 818 EUR
555-9075 555-9074 555-9072 555-9071
Capillary columns for dioxin and PCB analysis, TRACE™
Kolommen voor chromatografie
These columns are custom-tailored to analyse dioxins and polychlorinated biphenyls (PCBs). Optimised to meet the requirements of high-resolution GC/MS methods, these columns deliver optimum performance with exceptional stability and very low bleed. These low-polarity columns provide wide coverage of the 17 cogeners with the highest toxicological significance (2,3,7,8-chlorine substituted congeners).
  • Excellent quality and reproducibility
  • Low bleed, even at elevated temperatures
  • Outstanding robustness for difficult separations
  • Reliable and reproducible performance