Cover glasses
Leverancier: KNITTEL
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
VD11515Y100A VD12126Y100A VD12440Y100A VD12432Y100A VD12550Y100A VD12460Y100A
KNITVD11515Y100AEA 44.2 EUR
KNITVD11515Y100A KNITVD12126Y100A KNITVD12440Y100A KNITVD12432Y100A KNITVD12550Y100A KNITVD12460Y100A
Cover glasses
Dekglaasjes
Borosilicate glass of hydrolytic class 1 cover glasses with precise cutting.
Certificaten: ISO 8255/1
Verpakking: 100 pieces per box - 1000 pieces per carton.