Detectable plasters
Leverancier: SOEHNGEN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
1009986 1009981 1009984 1009982 1009988
SOEH1009986EA 0 EUR
SOEH1009986 SOEH1009981 SOEH1009984 SOEH1009982 SOEH1009988
Detectable plasters
Eerste-hulpbenodigdheden Wondverbanden
These blue, detectable plasters are specially designed for use in the food industry. Dispenser containing aluminium strips with an integrated metal surface that can be detected via a metal detector.
  • Plasters are detectable by most metal detectors used within the food industry
  • Breathable and skin-friendly
  • High quality hypoallergenic plasters