Multiroll tape dispensers, Write-On™, SP Bel-Art
Leverancier: Bel-Art Products, a Part of SP
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
F134720000 F134730000
BELAF134720000EA 413 EUR
BELAF134720000 BELAF134730000
Multiroll tape dispensers, Write-On™, SP Bel-Art
Etiketten- en tapedispensers
This dispenser provides a smooth writing surface, making labelling easy and fast. When marked tape is advanced and cut off on the serrated edge, a fresh length of label tape is positioned on the writing platform.
  • Smooth and snag free for up to four rolls of tape
  • Pen holder wells help to keep writing instruments handy
  • Spindles easily removed for loading and unloading, and adjusted to hold either 25 or 75 mm core diameter rolls
Leveringsinformatie: Incl. mounting holes and separating discs.