Laboratory jacks
Leverancier: JUCHHEIM Laborgerate GmbH
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
VL02280 VL02290 VL02291/3 VL02292 VL02293
JUCHVL02280EA 63.42 EUR
JUCHVL02280 JUCHVL02290 JUCHVL02291/3 JUCHVL02292 JUCHVL02293
Laboratory jacks
Heftafels voor laboratoria
Aluminium alloy, with epoxy resin coating. These manually operated lifting stages provide high stablity.
  • Special epoxy resin coating ensures that the surfaces are extremely resistant to chemical attack and to other environmental factors
  • Strict quality controls, as required by DIN EN ISO 9001:2000, ensure a constant product quality
  • Made in Germany