Ampoule breakers
Leverancier: Hach
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
2484600 2196800
HACH2484600EA 32.8 EUR
HACH2484600 HACH2196800
Ampoule breakers
Ampoule Openers
These breakers for ampoules are designed to prevent contamination. Breaker snaps ampoule tip off.