Gloves dispensers
Leverancier: Koehler Instrument
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
112-0099EA 121 EUR
112-0099 112-0098
Gloves dispensers
Handschoendispensers
These acrylic dispensers are suitable for any standard disposable glove box and protect the boxes from splashes.
  • Designed for wall mounting
  • Suitable for cleanroom applications
  • Boxes can be inserted from the side and gloves can be removed from the front