Storage Boxes, PP, with Lid
Leverancier: KOEHLER TECHNISCHE PRODUKTEN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
A-1199 A-1193 A-1188 A-1189 A-1192
212-9277EA 16.5 EUR
212-9277 212-3911 KOEHA-1188 KOEHA-1189 KOEHA-1192
Storage Boxes, PP, with Lid
Dozen Opbergdozen
Top storage boxes made of polypropylene (PP) for dust-free storage of tubes.
  • Holds 100 tubes up to 12 mm diameter
  • Temperature resistant from −40 to +130 °C