Cleanroom Waste Receptacles
Leverancier: Palbam
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
WR-3001 WR-3002 WR-3003 GW0501
134-0505EA 1140 EUR
134-0505 134-0506 129-1095 134-0509
Cleanroom Waste Receptacles
Bakken Afvalbakken
Stainless steel 304, electropolished finish.
  • No need to come in contact with the bin
  • No flaps or other devices are attached, as these just increase contact between user and the waste bin
  • Free-standing model is mounted on castors which are hidden from view
These round waste receptacles include an internal nylon holder and lid. The lid has a large opening for easy disposal.
Order Now
Specificaties
Environmentally Preferable