Culture tubes with screw cap
Leverancier: schuett-biotec GmbH
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
3563423 3560103 3562123 3562143 3563143 3561103
391-0157EA 227 EUR
391-0157 391-6135 SCUT3562123 SCUT3562143 SCUT3563143 SCUT3561103
Culture tubes with screw cap
Buisjes Cultuurbuisjes
Tubes made of borosilicate with aluminium or plastic screw caps and rubber seal.
  • Heavy walled
  • Easy to inoculate and clean