Blank flanges, type KF
Leverancier: Welch by Gardner Denver
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
701263 701261 701253 701251
ILMV701263EA 15.3 EUR
ILMV701263 ILMV701261 ILMV701253 ILMV701251
Blank flanges, type KF
Fittingen en connectoren
Blank flanges KF made of nickel-plated aluminium or stainless steel.
  • Reliable
  • Simple detachment for cleaning
Used for vacuum system.