External centering rings, type KF
Leverancier: Welch by Gardner Denver
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
701151 701143 701141
ILMV701151EA 8.9 EUR
ILMV701151 ILMV701143 ILMV701141
External centering rings, type KF
Afdichtingen Afdichtringen
External centering rings for double sided mechanism made of aluminium, with perbunan O-ring.