Centring rings with fine filter, type KF
Leverancier: Gardner Denver
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
ILMV701181EA 18 EUR
ILMV701181 ILMV701193 ILMV701183 181-0622
Centring rings with fine filter, type KF
Afdichtingen Afdichtringen
Centring rings made of stainless steel, with fine filter and perbunan O-ring.
Mesh width: 1,4 mm
Wire diameter: 0,2 mm