Ball check valves, manual, ABV series
Leverancier: Welch by Gardner Denver
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
700131 700132
ILMV700131EA 377.7 EUR
ILMV700131 ILMV700132
Ball check valves, manual, ABV series
Ventielen Vacuümventielen
These ball check valves have a PTFE hollow ball and viton seals.
  • Compact design
  • Housing made of aluminium