Butterfly valves, VK series
Leverancier: Welch by Gardner Denver
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
704523 704522
ILMV704523EA 429 EUR
ILMV704523 ILMV704522
Butterfly valves, VK series
Ventielen Vacuümventielen
These manually operated butterfly valves are made of stainless steel with viton seal.
Leak rate 10-7 mbar L/s
Order Now
Specificaties
Environmentally Preferable