Tissue processing cassette, Histosette II
Leverancier: DELTALAB
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
44140 44141 44143 44145 44147
631-9670EA 174 EUR
631-9670 631-9671 631-9672 631-9673 631-9674
Tissue processing cassette, Histosette II
Histologie-cassettes Cassettes voor weefselbehandeling
Made of acetal polymer.
  • With 45° angle writing area
  • Suitable for automatic labellers
  • No need to break the lid and then put it in place, as it is supplied pre-mounted
  • Can be easily opened and closed with just one hand