Hydrometers, Baumé
Leverancier: Amarell
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
H842400 H842476 H843198 H842402 H842814 H842878 H843148 H842478 H842474 H842876 H842358 H843154 H842480 H842404 H842818 H842810 H843196 H842350 H843100
614-0535EA 24.3 EUR
614-0535 614-0538 AMARH843198 AMARH842402 AMARH842814 AMARH842878 614-0536 614-0539 614-0541 AMARH842876 AMARH842358 AMARH843154 AMARH842480 AMARH842404 AMARH842818 AMARH842810 AMARH843196 AMARH842350 AMARH843100
Hydrometers, Baumé
Hydrometers
General purpose Baumé hydrometers with or without thermometer.
  • Reference temperature of 15 °C
  • Paper scale with visible print
  • Shot weighted body
Colourless round capillary with red or blue filling.