PCR tube racks with lid, 96-well
Leverancier: DELTALAB
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
M-553 M-550 M-551 M-552
DELTM-553EA 166 EUR
DELTM-553 DELTM-550 DELTM-551 DELTM-552
PCR tube racks with lid, 96-well
Rekken Tube Racks
Racks made of PP. Suitable for PCR individual tubes, 96 well plate and strips of 8 or 12 tubes.
  • Alphanumerical grid helps in sample identification
  • Complete with transparent lid
  • It can be used alone or inserted with workstation
  • Withstands temperature from −90 to 121 °C
Lid has a non-slip surface to allow safe storage of multiple racks.
W×D×H: 125×85×15 mm
Order Now
Specificaties
Environmentally Preferable