Hematocrit tubes, HEMATO-CLAD®
Leverancier: DRUMMOND
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
1-000-7500HC/5 1-000-7500-C/5
DRUM1-000-7500HC/5EA 227 EUR
DRUM1-000-7500HC/5 DRUM1-000-7500-C/5
Hematocrit tubes, HEMATO-CLAD®
Diagnostische systemen Clinical Sample Collection/Preparation Blood Sample Collection/Preparation
HEMATO-CLAD® hematocrit tubes combine the precision of glass with the safety provided by a Mylar overwrap.
  • Reduced contamination and accidental breakage
  • Mylar wrapping will provide an added layer of protection against cuts from broken glass