Jars, with pressure seal cap
Leverancier: LAMAPLAST
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
54 28 29
215-4060EA 3.8 EUR
215-4060 215-4062 215-4063
Jars, with pressure seal cap
Potjes Potjes voor algemeen gebruik
Jars, made of HDPE, with pressure sealing cap.
  • Pressure cap is tamper proof
  • Suitable for food storage
  • Good chemical and temperature resistance
Ideal for the storage of creams, semi-solid substances, granulates and powders.